missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human Trachea Membrane Tissue Lysate (Adult Membrane Normal)
Shop All Bio Techne ProductsDescription
The Human Trachea Membrane Tissue Lysate (Adult Membrane Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Respiratory |
| Quantity | 0.1 mg |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Membrane Protein - Human Adult Normal Tissue: Trachea |
| Cellular Fraction | Membrane |
| Tissue Condition | Normal |
| Cross Reactivity | Human |
| Host Species | Human |
| Concentration | 20 mg/mL |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?