missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TPTE2 (aa 269-291) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66988 (PA5-66988. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
TPIP is a member of a large class of membrane-associated phosphatases with substrate specificity for the 3-position phosphate of inositol phospholipids.[supplied by OMIM, Jul 2002].
Spécification
Spécification
| Accession Number | Q6XPS3 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 93492 |
| Name | Human TPTE2 (aa 269-291) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | Lipid phosphatase TPIP; phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase TPTE2; phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase TPTE2; PTEN-like inositol lipid phosphatase; TPIP; TPIP lipid phosphatase; TPTE and PTEN homologous inositol lipid phosphatase; TPTE2; transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 |
| Common Name | TPTE2 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu