missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TPT1/TCTP Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF4648
This item is not returnable.
View return policy
Description
TPT1/TCTP Polyclonal specifically detects TPT1/TCTP in Human samples. It is validated for Western Blot.
Specifications
| TPT1/TCTP | |
| Polyclonal | |
| Unconjugated | |
| P13693 | |
| TPT1 | |
| E. coli-derived recombinant human TPT1/TCTP Met1-Cys172 Accession # P13693 | |
| 100 μg | |
| Primary | |
| Detects human TPT1/TCTP in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse TPT1 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| FLJ27337, Fortilin, Histamine-releasing factor, HRF, p02, p23, TCTP, TCTPHRFfortilin, translationally-controlled tumor protein, tumor protein, translationally-controlled 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7178 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction