missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TPSD1 (aa 88-116) Control Fragment Recombinant Protein

Product Code. 30205880
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205880

Brand: Invitrogen™ RP104454

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (59%), Rat (59%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62007 (PA5-62007. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Tryptases comprise a family of trypsin-like serine proteases, the peptidase family S1. Tryptases are enzymatically active only as heparin-stabilized tetramers, and they are resistant to all known endogenous proteinase inhibitors. Several tryptase genes are clustered on chromosome 16p13.3. These genes are characterized by several distinct features. They have a highly conserved 3' UTR and contain tandem repeat sequences at the 5' flank and 3' UTR which are thought to play a role in regulation of the mRNA stability. Although this gene may be an exception, most of the tryptase genes have an intron immediately upstream of the initiator Met codon, which separates the site of transcription initiation from protein coding sequence. This feature is characteristic of tryptases but is unusual in other genes. Tryptases have been implicated as mediators in the pathogenesis of asthma and other allergic and inflammatory disorders. TPSD1 was once considered to be a pseudogene, although it is now believed to be a functional gene that encodes a protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BZJ3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23430
Name Human TPSD1 (aa 88-116) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Delta-tryptase; hmMCP-3-like tryptase III; Mast cell mMCP-7-like; mast cell tryptase; MCP7L1; MCP7-LIKE; MMCP-7 L; mMCP-7-like delta II tryptase; mMCP-7-like-1; mMCP-7-like-2; TPSD1; Tryptase delta; tryptase delta 1; tryptase-3
Common Name TPSD1
Gene Symbol TPSD1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KDLAALRVQLREQHLYYQDQLLPVSRIIV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.