missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TPP2 (aa 1029-1175) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65570 (PA5-65570. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain.
Spezifikation
Spezifikation
| Accession Number | P29144 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 7174 |
| Name | Human TPP2 (aa 1029-1175) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | TPP2; TPP-2; TppII; TPP-II; Tripeptidyl aminopeptidase; tripeptidyl peptidase 2; tripeptidyl peptidase II; tripeptidyl-peptidase 2; Tripeptidyl-peptidase II |
| Common Name | TPP2 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur