missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human TPO (Thrombopoietin) Recombinant Protein

Product Code. p-5232853
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11370842

Brand: Gibco™ PHC9511

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute lyophilized recombinant human Thrombopoietin in PBS containing 0.1% human serum albumin. Further dilutions should be made in low endotoxin medium or a buffered solution containing a carrier protein such as heat inactivated FCS or tissue culture grade BSA. Storage: Store this lyophilized preparation at <-20°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at <-20°C. Avoid repeated freeze/thaw cycles.

Thrombopoietin (TPO, THPO, MPL ligand, megakaryocyte colony-stimulating factor) is responsible for the generation of platelets through its action on hematopoietic cells committed to the megakaryocyte lineage. Binding of thrombopoietin to its receptor, MPL (CD110), initiates a signaling cascade via JAK/STAT, MAPK, and PI3K/AKT signaling pathways that leads to the proliferation and differentiation of megakaryocytes and, ultimately, to the production of platelets (thrombopoiesis). Thrombopoietin is constitutively produced by the liver and has a predicted molecular weight of 38 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P40225
For Use With (Application) Bioactivity
Formulation PBS with no preservative
Gene ID (Entrez) 7066
Molecular Weight (g/mol) 85 kDa
Name Human TPO (Thrombopoietin)
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias C-mpl ligand; Megakaryocyte colony-stimulating factor; megakaryocyte growth and development factor; megakaryocyte stimulating factor; Mgdf; MKCSF; ML; MPL ligand; Mpllg; MSA; Myeloproliferative leukemia virus oncogene ligand; prepro-thrombopoietin; TDH2A; THCYT1; THPO; Thrombopoietin; thyroid microsomal antigen; thyroid peroxidase; thyroperoxidase; Tpo; TPX
Common Name Thrombopoietin
Gene Symbol THPO
Biological Activity ED50 = 0.5 - 5 ng/mL; determined by dose-dependent proliferation of human TF-1 cells.
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Content And Storage -20°C
Classification Carrier-Free
Activity ED50 = 0.5 - 5 ng/mL; determined by dose-dependent proliferation of human TF-1 cells.
Bioassay Proliferation of TF1 cells
Endotoxin Level <1 EU/ml
Shipping Condition Dry Ice
Expression System HEK293 cells
Protein Family Growth Factors & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype TPO
Research Category Endocrinology, Stem Cell Research, Angiogenesis
Product Line Gibco
Purity or Quality Grade >90% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.