missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TOX2 (aa 35-85) Control Fragment Recombinant Protein

Product Code. 30180486
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30180486

Brand: Invitrogen™ RP99344

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TOX2 (TOX High Mobility Group Box Family Member 2) is a Protein Coding gene. Diseases associated with TOX2 include Conduct Disorder. Gene Ontology (GO) annotations related to this gene include chromatin binding and RNA polymerase II transcription factor binding. An important paralog of this gene is TOX.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96NM4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84969
Name Human TOX2 (aa 35-85) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI851523; AV026525; BI987407; C20orf100; dJ1108D11.2; dJ495O3.1; GCX1; Gcx-1; granulosa cell HMG box 1; granulosa cell HMG box protein 1; granulosa cell HMG-box protein 1; granulosa cell HMG-box protein-1; LOW QUALITY PROTEIN: TOX high mobility group box family member 2; RxHMG1; TOX high mobility group box family member 2; TOX2
Common Name TOX2
Gene Symbol TOX2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IAGVFPQKFDGDSAYVGMSDGNPELLSTSQTYNGQSENNEDYEIPPITPPN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.