missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total p27/Kip1 DuoSet IC, R&D Systems™
Human Total p27/Kip1 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 62.50 to 4000 pg/mL |
---|---|
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Gene ID (Entrez) | 1027 |
Product Type | ELISA Development Kit for intracellular proteins |
Conjugate | Biotin |
Description
The Human Total p27/Kip1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human p27/Kip1.Specifications
62.50 to 4000 pg/mL | |
1027 | |
Biotin |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
ELISA Development Kit for intracellular proteins | |
Human |