missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Total MMP-14/MT1-MMP DuoSet ELISA

Product Code. 18745457
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 18745457

Brand: R&D Systems™ DY91805

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Total MMP-14/MT1-MMP DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Total MMP-14/MT1-MMP DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human MMP-14/MT1-MMP.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.63 to 20 ng/mL
Conjugate Biotin
Immunoassay Kit Format 96-well Strip Plate
Product Type Solid Phase Sandwich ELISA
Sufficient For Five 96-well plates
For Use With (Application) ELISA
Gene ID (Entrez) 4323
Gene Symbol MMP14
Quantity 1 Kit (for 5 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP14, MMP-14, MMP- x 1, MT1MMP, MT1-MMP, MT1-MMPMTMMP1, MT-MMP 1, MT-MMP1
Target MMP-14/MT1-MMP
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.