missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total EphA2 DuoSet IC, R&D Systems™
contains the basic components required for the development of sandwich ELISAs to measure in cell lysates
POA
Specifications
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
---|---|
Gene ID (Entrez) | 1969 |
Product Type | ELISA Development Kit for intracellular proteins |
Conjugate | Biotin |
Species | Human |
Description
- Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
- Development protocols are provided to guide further assay optimization
- Assay can be customized to your specific needs
Specifications
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
ELISA Development Kit for intracellular proteins | |
Human |
1969 | |
Biotin |