missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total Bax-alpha DuoSet IC ELISA
missing translation for 'orderingAttributeHoverText'
Packaging Quantity:
2 Plates
5 Plates
missing translation for 'unitSize'
1 set
Description
Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously.
The Human Total Bax-alpha DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human Bax-alpha.
Specifications
Specifications
| Product Type | ELISA Development Kit for Intracellular Proteins |
| Quantity | 1 Kit |
| Species | Human |
| Conjugate | Biotin |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 581 |
| Packaging Quantity | 5 Plates |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu