missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TOR1AIP2 (aa 4-80) Control Fragment Recombinant Protein

Product Code. 30201503
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201503

Brand: Invitrogen™ RP107753

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (48%), Rat (48%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67110 (PA5-67110. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

One of the two protein isoforms encoded by this gene is a type II integral membrane protein found in the endoplasmic reticulum (ER). The encoded protein is a cofactor for the ATPase TorsinA, regulating the amount of TorsinA present in the ER compared to that found in the nuclear envelope. Defects in this protein are a cause of early onset primary dystonia, a neuromuscular disease. The other isoform encoded by this gene is an interferon alpha responsive protein whose cellular role has yet to be determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NFQ8, Q9H496
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 163590
Name Human TOR1AIP2 (aa 4-80) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110020D10Rik; 15 kDa interferon-responsive protein; 15 kDa; A130072J07; AA103493; AW060462; AW610675; C77739; Ifrg15; interferon alpha responsive gene, 15 kDa; interferon alpha responsive protein (15 kDa); interferon alpha responsive protein; torsin-1 A-interacting protein 2; interferon responsive gene 15; Lull1; lumenal domain like LAP1; lumenal domain-like LAP1; NET9; RP11-12M5.5; TOR1AIP2; torsin 1 A interacting protein 2; torsin A interacting protein 2; torsin-1 A-interacting protein 2; Torsin-1 A-interacting protein 2, isoform IFRG15
Common Name TOR1AIP2
Gene Symbol TOR1AIP2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SGLREPQEDSQKDLENDPSVNSQAQETTIIASNAEEAEILHSACGLSKDHQEVETEGPESADTGDKSESPDEANVGK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.