missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TNRC6B (aa 1274-1401) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51849 (PA5-51849. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The function of TNRC6B remains unknown.
Specifications
Specifications
| Accession Number | Q9UPQ9 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 23112 |
| Name | Human TNRC6B (aa 1274-1401) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2700090M07Rik; A730065C02Rik; AI848765; androgen receptor-related apoptosis-associated protein CBL27; Cbl27; D230019K20Rik; KIAA1093; TNRC6B; trinucleotide repeat containing 6 B; trinucleotide repeat containing adaptor 6 B; trinucleotide repeat-containing gene 6 B protein |
| Common Name | TNRC6B |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction