missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TNNT1 (aa 21-59) Control Fragment Recombinant Protein

Product Code. 30203816
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30203816

Brand: Invitrogen™ RP103332

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84381 (PA5-84381. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein that is a subunit of troponin, which is a regulatory complex located on the thin filament of the sarcomere. This complex regulates striated muscle contraction in response to fluctuations in intracellular calcium concentration. This complex is composed of three subunits: troponin C, which binds calcium, troponin T, which binds tropomyosin, and troponin I, which is an inhibitory subunit. This protein is the slow skeletal troponin T subunit. Mutations in this gene cause nemaline myopathy type 5, also known as Amish nemaline myopathy, a neuromuscular disorder characterized by muscle weakness and rod-shaped, or nemaline, inclusions in skeletal muscle fibers which affects infants, resulting in death due to respiratory insufficiency, usually in the second year. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13805
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 7138
Name Human TNNT1 (aa 21-59) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias ANM; AW146156; Fang2; MGC104241; NEM5; skeletal muscle slow-twitch TnT; slow skeletal muscle troponin T; ssTnT; sTnT; Tnnt1; TNT; TNTS; troponin T slow skeletal muscle type; troponin T slow type isoform sTnT1; troponin T slow type isoform sTnT2; troponin T type 1 (skeletal, slow); troponin T, slow skeletal muscle; troponin T1, skeletal, slow; troponin T1, slow skeletal type; troponin-T1, skeletal, slow; zgc:193831; zgc:193865
Common Name TNNT1
Gene Symbol TNNT1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EEEEAPEEPEPVAEPEEERPKPSRPVVPPLIPPKIPEGE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.