missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TNIP2 (aa 221-298) Control Fragment Recombinant Protein

Product Code. 30205007
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205007

Brand: Invitrogen™ RP104131

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64096 (PA5-64096. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TNFAIP3, also known as A20, is located in chromosome band 6q23, a region that is often deleted in B cell lymphomas. Recently, it was identified as a tumor suppressor gene in Hodgkin lymphoma and several subtypes of non-Hodgkin lymphomas. TNFAIP3 was initially identified as a zinc-finger protein that is rapidly and transiently induced by TNF-a, inhibiting NF-kappa-B-dependent gene expression, and protecting cells from TNF-a-cytotoxicity. Overexpression of TNFAIP3 also inhibits the TLR2- and TLR4-mediated interleukin-8 synthesis in airway epithelial cells, suggesting that TNFAIP3 also acts as a negative regulator of TLR-mediated inflammatory responses, thereby protecting the host against harmful over-responses to pathogens. At least two isoforms of TNFAIP3 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NFZ5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79155
Name Human TNIP2 (aa 221-298) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1810020H16Rik; A20 binding inhibitor of NF-kappaB activation-2; A20-binding inhibitor of NF-kappa-B activation 2; A20-binding inhibitor of NF-kappaB activation-2; ABIN2; ABIN-2; AI428870; Fetal liver LKB1-interacting protein; FLIP1; KLIP; TNFAIP3 interacting protein 2; TNFAIP3-interacting protein 2; Tnip2
Common Name TNIP2
Gene Symbol Tnip2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EDLNAKWQRYNASRDEYVRGLHAQLRGLQIPHEPELMRKEISRLNRQLEEKINDCAEVKQELAASRTARDAALERVQM
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.