missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TNF RII/TNFRSF1B Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems MAB226-SP
This item is not returnable.
View return policy
Description
TNF RII/TNFRSF1B Monoclonal specifically detects TNF RII/TNFRSF1B in Human samples. It is validated for Flow Cytometry, Neutralization, Immunocytochemistry.
Specifications
| TNF RII/TNFRSF1B | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNFRII, TNF-RII, TNF-R-II, TNFR-II, TNFRSF1B, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 7133 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry, Neutralization, Immunocytochemistry | |
| 22221 | |
| Flow Cytometry 0.25 ug/10^6 cells, Neutralization 0.5-1.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P20333 | |
| TNFRSF1B | |
| E. coli-derived recombinant human TNF RII Leu23-Asp257 Accession # P20333 | |
| 25 μg | |
| Primary | |
| Detects human TNF RII/TNFRSF1B in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) 4-1BB, rhCD27, rhCD30, rhCD40, rhDR3, rhDR6, rhFas, rhGITR, rhHVEM, rhLTR beta , rhNGF R, rhOPG, rhRANK, rhTNF RI, or recombinant mouse TNF RII is observed. Neutralizes the biological effect of rhTNF RII but not rhTNF RI. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction