missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human TNF-RII (soluble) Recombinant Protein, PeproTech®

Product Code. 17832213
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17832213

Brand: Gibco™ 3101220UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human sTNF Receptor Type II is an 18.9 kDa protein (174 amino acid residues) comprising the cysteine-rich, ligand binding portion of the extracellular domain of the TNFRII protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TNFR2 (tumor necrosis factor receptor superfamily member 1B) is a receptor with high affinity for TNFSF2/TNF-alpha. It has a much lower affinity for homotrimeric TNFSF1/lymphotoxin-alpha. The TRAF1/TRAF2 complex recruits the apoptotic suppressors BIRC2 and BIRC3 to TNFRSF1B/TNFR2. TNFR2 mediates most of the metabolic effects of TNF-alpha.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P20333
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 7133
Molecular Weight (g/mol) 18.9 kDa
Name Human TNF-RII (soluble)
Quantity 20 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CD120b; Etanercept; p75; p75 TNF receptor; p75 TNFR; p75TNFR; p80 TNF-alpha receptor; sCD120b; soluble CD120b; soluble TNFR1B variant 1; soluble TNFR2; sTNF R2; sTNFR 2; sTNFR2; TBP-2; TBPII; TNF receptor beta chain; TNF receptor superfamily member 1 B; TNF-alphaR2; TNFalpha-R2; TNFBR; TNFR; TNFR II; Tnfr-1; TNFR1B; TNFR2; TNF-R2; Tnfr-2; TNF-R75; TNFR80; TNFRII; TNF-RII; TNF-R-II; TNFR-II; TNFRSF1B; tumor necrosis factor beta receptor; tumor necrosis factor binding protein 2; Tumor necrosis factor receptor; tumor necrosis factor receptor 2; Tumor necrosis factor receptor superfamily member 1 B; Tumor necrosis factor receptor superfamily member 1 b, membrane form; tumor necrosis factor receptor superfamily, member 1 B; tumor necrosis factor receptor type II; Tumor necrosis factor-binding protein 2
Common Name TNFR2
Gene Symbol Tnfrsf1b
Biological Activity Determined by its inhibitory effect of the TNF-alpha mediated cytotoxicity in murine L-929 cells. The ED50 for this effect in the presence of 0.25 ng/ml of Recombinant Human TNF-alpha, is 0.125 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence MAPEPGSTCR LREYYDQTAQ MCCSKCSPGQ HAKVFCTKTS DTVCDSCEDS TYTQLWNWVP ECLSCGSRCS SDQVETQACT REQNRICTCR PGWYCALSKQ EGCRLCAPLR KCRPGFGVAR PGTETSDVVC KPCAPGTFSN TTSSTDICRP HQICNVVAIP GNASMDAVCT STSP
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.