missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TNF-RII (soluble) Recombinant Protein
Description
RP-75727 contains purified recombinant, soluble human TNF Receptor-2 protein with an N-terminal hexahistidine tag. RP-75727 may be used as a positive control in many immunological assays.
TNFR2 (tumor necrosis factor receptor superfamily member 1B) is a receptor with high affinity for TNFSF2/TNF-alpha. It has a much lower affinity for homotrimeric TNFSF1/lymphotoxin-alpha. The TRAF1/TRAF2 complex recruits the apoptotic suppressors BIRC2 and BIRC3 to TNFRSF1B/TNFR2. TNFR2 mediates most of the metabolic effects of TNF-alpha.
Specifications
Specifications
| Accession Number | P20333 |
| Concentration | 0.3 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | tris HCl with 50% glycerol, 5 mM EDTA and no preservative; pH 8 |
| Gene ID (Entrez) | 7133 |
| Name | Human TNF-RII (soluble) |
| pH Range | 8 |
| Quantity | 10 μg |
| Source | E. coli |
| Immunogen | N/A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?