missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human TNF RI/TNFRSF1A Quantikine QuicKit ELISA
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 96
Specifications
Specifications
| Assay Range | 15.60 to 1000 pg/mL |
| Assay Sensitivity | 1.69 pg/mL |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Heparin Plasma, Serum, Urine |
| Specificity | Natural and Recombinant Human TNF RI. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 7132 |
| Host Species | Human |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2 to 8 °C. Do not use past expiration date |
| Synonym | CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNFRI, TNF-RI, TNF-R-I, TNFR-I, TNFRSF1A, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1 A isoform beta, tumor necrosis factor receptor superfamily member 1 A, tumor necrosis factor receptor superfamily, member 1 A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction