missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human TNF RI/TNFRSF1A DuoSet ELISA
Click to view available options
Quantity:
5 Plates
Unit Size:
1 set
Description
Includes
Human TNF RI Capture Antibody; Human TNF RI Detection Antibody; Human TNF RI Standard; Streptavidin-HRP
The Human TNF RI/TNFRSF1A DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human TNF RI/TNFRSF1A.
Specifications
Specifications
| Quantity | 5 Plates |
| Species | Human |
| Conjugate | Biotin |
| For Use With (Application) | ELISA |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction