missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Lymphotoxin-alpha/TNF-beta DuoSet ELISA

Product Code. p-4754512
Click to view available options
Format:
15 96-well Plates
5 96-well Plates
Unit Size:
1 set
Pack of 5
This item is not returnable. View return policy

Product Code. 30021467

Brand: R&D Systems™ DY211

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Lymphotoxin-alpha/TNF-beta DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Lymphotoxin-alpha/TNF-beta DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Lymphotoxin-alpha/TNF-beta.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Kit
Quantity 1 Kit (for 15 Plates)
Species Human
Conjugate Biotin
Assay Range 31.20 to 2000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Format 15 96-well Plates
For Use With (Application) ELISA
Gene ID (Entrez) 4049
Synonym LT, LTA, LT-alpha, Lymphotoxin alpha, lymphotoxin alpha (TNF superfamily, member 1), Lymphotoxinalpha, tnfb, TNF-beta, TNFBlymphotoxin-alpha, TNFSF1, TNFSF1B, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.