missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Lymphotoxin-alpha/TNF-beta DuoSet ELISA
Click to view available options
Format:
15 96-well Plates
5 96-well Plates
Unit Size:
1 set
Pack of 5
Description
The Human Lymphotoxin-alpha/TNF-beta DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Lymphotoxin-alpha/TNF-beta.
Specifications
Specifications
| Product Type | ELISA Kit |
| Quantity | 1 Kit (for 15 Plates) |
| Species | Human |
| Conjugate | Biotin |
| Assay Range | 31.20 to 2000 pg/mL |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| Format | 15 96-well Plates |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 4049 |
| Synonym | LT, LTA, LT-alpha, Lymphotoxin alpha, lymphotoxin alpha (TNF superfamily, member 1), Lymphotoxinalpha, tnfb, TNF-beta, TNFBlymphotoxin-alpha, TNFSF1, TNFSF1B, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction