missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TNF-beta Recombinant Protein

Product Code. 11569051
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity unitSize
11569051 1 mg 1mg
1 options
This item is not returnable. View return policy

Product Code. 11569051

Brand: Invitrogen™ RP75741

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

RP-75741 contains purified recombinant human TNF beta protein with an N-terminal hexahistidine tag. RP-75741 may be used as a positive control in many immunological assays.

TNF beta, a member of the tumor necrosis factor family, is a cytokine produced by lymphocytes. The protein is highly inducible, secreted, and forms heterotrimers with lymphotoxin-beta which anchor lymphotoxin-alpha to the cell surface. This protein also mediates a large variety of inflammatory, immunostimulatory, and antiviral responses, is involved in the formation of secondary lymphoid organs during development and plays a role in apoptosis. Genetic variations in this gene are associated with susceptibility to leprosy type 4 and psoriatic arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01374
For Use With (Application) Control, Western Blot
Formulation 10 mM Bis-tris propane with 50% glycerol, 62.5 mM NaCl and no preservative; pH 9
Gene ID (Entrez) 4049
Name Human TNF-beta
Quantity 1 mg
Source E. Coli
Regulatory Status RUO
Gene Alias DAMA-25N12.13; hlb382; hypothetical protein; LT; LT[a]; LT-[a]; LTA; LTalpha; LT-alpha; Ltx; lymphotoxin A; lymphotoxin alpha; lymphotoxin alpha (TNF superfamily, member 1); lymphotoxin-alpha; lymphotoxin-alpha; lymphotoxin alpha; TNF b; TNF beta; TNF superfamily, member 1; TNF β; Tnfb; TNFbeta; TNF-beta; TNF-N; Tnfsf1; Tnfsf1b; TNFβ; TNLG1E; tumor necrosis factor beta; tumor necrosis factor ligand 1 E; tumor necrosis factor ligand superfamily member 1
Common Name TNF beta
Gene Symbol LTA
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Leu35-Leu205 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.