Learn More
R&D Systems™ Human TNF-alpha ELISpot Development Module, 5 Plate
Product Code. 18412020
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 18412020
Brand: R&D Systems™ SEL210
Sufficient for at least five 96-well plates
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Synonym APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.