Learn More
Invitrogen™ Human TNF alpha ELISA Kits
Description
Assay TNFα (human tumor necrosis factor alpha) levels in human serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with our Invitrgen™ Human TNF alpha ELISA Kits.
Target: Natural and recombinant human TNFα (human tumor necrosis factor alpha)
Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement: Absorbance at 450nm minus absorbance at 550nm
Assay Range: 15.6 to 1000pg/mL
Sensitivity: < 2pg/mL
Precision: Dependent upon sample type and level (see instructions)
Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Total Assay Time: 3.5 hours
Sample Size: 50μL
Specificity: No significant cross-reaction with human IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12, IL-12p40, IL-13, IL-15, IL-16, IL-17, IL-18, TNFβ, Eotaxin, RANTES, GROα, GROβ, MCP-1, MCP-2, MCP-3, MCP-4, VEGF, GCSF, GMCSF, MIP-1α, MIP-1β, TGFβ, IFNγ, IFNα; mouse TNFα; rat TNFα; and bovine TNFα. TNFα Receptor types 1 and 2 tested at 40mg/mL did not interfere in this assay. Recombinant pig TNFα cross-reacted at 100%
Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Reagent Kit (previously called a “Screening Set”) contains matched antibodies, standard and detection reagents but no coated plates. The Antibody Pair Kit (previously called a “MiniKit”) contains only matched antibodies and standard (no plates or detection reagents).
Specifications
Specifications
| Assay Range | 15.6 to 1000pg/mL |
| Assay Sensitivity | <2pg/mL |
| Description | Human TNF alpha ELISA Reagent Kit |
| Immunoassay Kit Format | Colorimetric ELISA (TMB substrate) with Plates |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
| Specificity | No significant cross-reaction with human IL-1a, IL-1ß, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12, IL-12p40, IL-13, IL-15, IL-16, IL-17, IL-18, TNFß, Eotaxin, RANTES, GROa, GROß, MCP-1, MCP-2, MCP-3, MCP-4, VEGF, GCSF, GMCSF, MIP-1a, MIP |
| Sufficient For | 5 x 96 microplate wells |
| Kit Contents | Streptavidin-HRP Conjugate, 0.25mL, TNF alpha Detection Antibody, 1 vial, TMB Substrate Solution, 55mL, Stop Solution, 55mL, Human TNF alpha Standard, 5 vials, TNF alpha Coating Antibody, 1 vial |
| Measurement | Absorbance at 450nm minus absorbance at 550nm |
| Precision | Dependent upon sample type and level (see instructions) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.