missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TMPRSS2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£170.00 - £416.00
Specifications
| Antigen | TMPRSS2 |
|---|---|
| Clone | 1038105 |
| Dilution | Flow Cytometry 0.25 ug/106 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18713757
![]() |
R&D Systems
MAB107231-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18790527
![]() |
R&D Systems
MAB107231-100 |
100 μg |
£416.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMPRSS2 Monoclonal antibody specifically detects TMPRSS2 in Human samples. It is validated for Flow CytometrySpecifications
| TMPRSS2 | |
| Flow Cytometry 0.25 ug/106 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EC 3.4.21, Epitheliasin, FLJ41954, NP_001128571.1, NP_001369649.1, NP_005647.3, PP9284, PRSS10, Serine protease 10, TMPRSS2, transmembrane protease serine 2, transmembrane protease, serine 2 | |
| Human TMPRSS2 synthetic peptide, Accession # O15393 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1038105 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| O15393 | |
| 7113 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title