missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TMEM8A (aa 174-256) Control Fragment Recombinant Protein

Product Code. 30208885
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30208885

Brand: Invitrogen™ RP101281

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (73%), Rat (73%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62365 (PA5-62365. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Involved in the lipid remodeling steps of GPI-anchor maturation. Lipid remodeling steps consist in the generation of 2 saturated fatty chains at the sn-2 position of GPI-anchor proteins (GPI-AP). Has phospholipase A2 activity that removes an acyl-chain at the sn-2 position of GPI-anchors during the remodeling of GPI. Required for the shedding of the GPI-AP TDGF1, but not CFC1, at the cell surface. Shedding of TDGF1 modulates Nodal signaling by allowing soluble TDGF1 to act as a Nodal coreceptor on other cells (PubMed:27881714). Also indirectly involved in the translocation of RAC1 from the cytosol to the plasma membrane by maintaining the steady state amount of CAV1-enriched plasma membrane subdomains, stabilizing RAC1 at the plasma membrane (PubMed:27835684). In contrast to myomaker (TMEM8C), has no fusogenic activity (PubMed:26858401). [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9HCN3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 58986
Name Human TMEM8A (aa 174-256) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias five-span transmembrane protein M83; GPI processing phospholipase A2; GPI-PLA2; M83; PGAP6; Post-GPI attachment to proteins factor 6; Protein M83; TMEM6; TMEM8; TMEM8A; Transmembrane protein 6; Transmembrane protein 8; transmembrane protein 8 (five membrane-spanning domains); transmembrane protein 8 A
Common Name TMEM8A
Gene Symbol TMEM8A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TCAYVFQPELLVTRVVEISIMEPDVPLPQTLLSHPSYLKVFVPDYTRELLLELRDCVSNGSLGCPVRLTVGPVTLPSNFQKVL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.