Learn More
Invitrogen™ Human TMEM66 (aa 199-251) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65267 (PA5-65267. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q96BY9 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 51669 |
| Name | Human TMEM66 (aa 199-251) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1810045K07Rik; arrestin-E; FOAP-7; HBV XAg-transactivated protein 3; HBV X-transactivated gene 3 protein; HSPC035; NPD003; Protein FOAP-7; PSEC0019; Saraf; SARAF long isoform; SARAF short isoform; SOCE-associated regulatory factor; store-operated calcium entry associated regulatory factor; store-operated calcium entry-associated regulatory factor; testicular secretory protein Li 59; TMEM66; Transmembrane protein 66; UNQ1967/PRO4499; XTP3 |
| Common Name | TMEM66 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.