missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TMEM2 (aa 2-70) Control Fragment Recombinant Protein

Product Code. 30212613
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212613

Brand: Invitrogen™ RP90227

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (71%), Rat (71%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a type II transmembrane protein that belongs to the interferon-induced transmembrane (IFITM) protein superfamily. The encoded protein functions as a cell surface hyaluronidase that cleaves extracellular high molecular weight hyaluronan into intermediate size fragments before internalization and degradation in the lysosome. It also has an interferon-mediated antiviral function in humans through activation of the JAK STAT signaling pathway. The activation of this gene by transcription factor SOX4 in breast cancer cells has been shown to mediate the pathological effects of SOX4 on cancer progression. Naturally occurring mutations in this gene are associated with autosomal recessive non-syndromic hearing loss.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UHN6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23670
Name Human TMEM2 (aa 2-70) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias basic helix-loop-helix domain containing, class B, 2; BHLHB2; cell migration inducing hyaluronidase 2; Cell migration-inducing hyaluronidase 2; Cell surface hyaluronidase; CEMIP2; Kiaa1412; Tmem2; transmembrane protein 2
Common Name TMEM2
Gene Symbol CEMIP2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence YATDSRGHSPAFLQPQNGNSRHPSGYVPGKVVPLRPPPPPKSQASAKFTSIRREDRATFAFSPEEQQAQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.