missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TMEM184B (aa 343-407) Control Fragment Recombinant Protein

Product Code. 30193581
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30193581

Brand: Invitrogen™ RP92365

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54973 (PA5-54973. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TMEM184B, also known as C22orf5, is a 407 amino acid multi-pass membrane protein and represents a novel gene in the activation of the MAPK signaling pathway. The gene encoding TMEM184B maps to human chromosome 22; mutations in several of the genes in chromosome 22 are involved in the development of autism, schizophrenia, Phelan-McDermid syndrome and Neurofibromatosis type 2, suggesting that TMEM184B may play a role in these syndromes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y519
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 25829
Name Human TMEM184B (aa 343-407) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2610507A11; 4732495E13Rik; C22orf5; CTA-447C4.2; FM08; HS5O6A; HSPC256; PSEC0108; Putative MAPK-activating protein FM08; RGD1306591; Tmem184b; Transmembrane protein 184 B
Common Name TMEM184B
Gene Symbol TMEM184B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TMNPHDIVQDAIHNFSPAYQQYTQQSTLEPGPTWRGGAHGLSRSHSLSGARDNEKTLLLSSDDEF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.