missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TMEM151B (aa 131-171) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63077 (PA5-63077. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
TMEM151B is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.
Specifications
Specifications
| Accession Number | Q8IW70 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 441151 |
| Name | Human TMEM151B (aa 131-171) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | bA444E17.5; C6orf137; Gm323; TMEM151B; TMEM193; transmembrane protein 151 B; Transmembrane protein 193 |
| Common Name | TMEM151B |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction