Learn More
Invitrogen™ Human TMCO3 (aa 374-428) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61660 (PA5-61660. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q6UWJ1 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 55002 |
| Name | Human TMCO3 (aa 374-428) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | B230339H12Rik; C13orf11; C87304; LOW QUALITY PROTEIN: transmembrane and coiled-coil domain-containing protein 3; putative LAG1-interacting protein; RGD1306586; testis tissue sperm-binding protein Li 36 A; tmco3; Transmembrane and coiled-coil domain-containing protein 3; transmembrane and coiled-coil domain-containing protein 3; LOW QUALITY PROTEIN: transmembrane and coiled-coil domain-containing protein 3; transmembrane and coiled-coil domains 3; UNQ2419/PRO4976 |
| Common Name | TMCO3 |
| Show More |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.