missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TMBIM1 (aa 25-97) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52909 (PA5-52909. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Negatively regulates aortic matrix metalloproteinase-9 (MMP9) production and may play a protective role in vascular remodeling.
Specifications
Specifications
| Accession Number | Q969X1 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 64114 |
| Name | Human TMBIM1 (aa 25-97) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2310061B02Rik; AA960455; AU024746; C78899; Lfg3; mKIAA4161; MST100; MSTP100; PP1201; Protein lifeguard 3; Protein RECS1; Protein RECS1 homolog; PSEC0158; RECS1; Responsive to centrifugal force and shear stress gene 1 protein; Tmbib1; Tmbim1; transmembrane BAX inhibitor motif containing 1; transmembrane BAX inhibitor motif-containing protein 1 |
| Common Name | TMBIM1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction