missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The Human TLR9 ELISA Kit (Chemiluminescence) measures human TLR9 in serum, plasma and other biological fluids.
Specifications
Specifications
| Assay Range | 31.25 to 2000 pg/mL |
| Assay Sensitivity | 18.75 pg/mL |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Human TLR9/CD289 in samples. No significant cross-reactivity or interference between Human TLR9/CD289 and analogues was observed. |
| Gene ID (Entrez) | 54106 |
| Gene Symbol | TLR9 |
| Host Species | Human |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?