missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TLR9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3658
This item is not returnable.
View return policy
Description
TLR9 Monoclonal specifically detects TLR9 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| TLR9 | |
| Monoclonal | |
| LYOPH | |
| Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| NP_059138 | |
| TLR9 | |
| NS0 mouse myeloma cell line transfected with human TLR9 Accession # NP_059138 | |
| 100 μg | |
| Primary | |
| Detects human TLR9. Stains human TLR9-transfected cells but not irrelevant transfectants. | |
| Human | |
| IgM |
| Flow Cytometry, CyTOF | |
| 229106 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD289, CD289 antigen, toll-like receptor 9 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 54106 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction