missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TLR8 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC8999T-025
This item is not returnable.
View return policy
Description
TLR8 Monoclonal specifically detects TLR8 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| TLR8 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51311 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 935166 | |
| Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
| Q9NR97 | |
| TLR8 | |
| Chinese hamster ovary cell line CHO-derived recombinant human TLR8 Glu27-Thr827 Accession # Q9NR97 | |
| 25 Tests | |
| Primary | |
| Detects human TLR8 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction