missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TLR4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 17 publications
Brand: R&D Systems AF1478
This item is not returnable.
View return policy
Description
TLR4 Polyclonal specifically detects TLR4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF-ready.
Specifications
| TLR4 | |
| Polyclonal | |
| Unconjugated | |
| O00206 | |
| TLR4 | |
| Mouse myeloma cell line NS0-derived recombinant human TLR4 Glu24-Lys631 Accession # O00206 | |
| 100 μg | |
| Primary | |
| Detects human TLR4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant mouse (rm) TLR4 is observed and less than 5% cross-reactivity with recombinant human (rh) TLR1, rhTLR2, rhTLR3 and rmTLR6 is observed. | |
| Human | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Flow Cytometry 2.5 μg/10^6 cells, Immunohistochemistry 5-15 μg/mL, Neutralization 1.5-7.5 μg/mL, Immunocytochemistry 5-15 μg/mL, CyTOF-ready | |
| ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7099 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction