missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TLR4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB1478-SP
This item is not returnable.
View return policy
Description
TLR4 Monoclonal specifically detects TLR4 in Human samples. It is validated for Western Blot.
Specifications
| TLR4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O00206 | |
| TLR4 | |
| Mouse myeloma cell line NS0-derived recombinant human TLR4 Glu24-Lys631 Accession # O00206 | |
| 25 μg | |
| Primary | |
| Detects human TLR4 in direct ELISAs and Western blots. Shows approximately 25% cross-reactivity with recombinant mouse (rm) TLR4 and no cross-reactivity with recombinant human TLR1, 2, 3, or rmTLR6. | |
| Human | |
| Purified |
| Western Blot | |
| 285227 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7099 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction