missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Tissue alpha-L-Fucosidase/FUCA1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB7039
This item is not returnable.
View return policy
Description
Tissue alpha-L-Fucosidase/FUCA1 Monoclonal specifically detects Tissue alpha-L-Fucosidase/FUCA1 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| Tissue alpha-L-Fucosidase/FUCA1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Simple Western 2 ug/mL | |
| P04066 | |
| FUCA1 | |
| Mouse myeloma cell line NS0-derived recombinant human Tissue alpha-L-Fucosidase/FUCA1 Gln32-Lys466 Accession # P04066 | |
| 100 μg | |
| Primary | |
| Detects human Tissue alpha-L-Fucosidase/FUCA1 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 810607 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Alpha-L-fucosidase 1, Alpha-L-fucosidase I, Alpha-L-fucoside fucohydrolase 1, EC 3.2.1, EC 3.2.1.51, FUCA, FUCA1, fucosidase, alpha-L- 1, tissue, tissue alpha-L-fucosidase, Tissue alphaLFucosidase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2517 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction