missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TIMP-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB973-100
This item is not returnable.
View return policy
Description
TIMP-3 Monoclonal specifically detects TIMP-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, ELISA Capture (Matched Antibody Pair).
Specifications
| TIMP-3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HSMRK222, K222, K222TA2, metalloproteinase inhibitor 3, MIG-5 protein, Protein MIG-5, pseudoinflammatory), SFD, TIMP metallopeptidase inhibitor 3, TIMP3, TIMP-3, tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, Tissue inhibitor of metalloproteinases 3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7078 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunoprecipitation, ELISA | |
| 183551 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P35625 | |
| TIMP3 | |
| Mouse myeloma cell line NS0-derived recombinant human TIMP-3 Cys24-Pro211 Accession # P35625 | |
| 100 μg | |
| Primary | |
| Detects human TIMP-3 in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human TIMP-1, -2, or -4 is observed. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction