missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TIMP-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 15 publications
Brand: R&D Systems MAB970-SP
This item is not returnable.
View return policy
Description
TIMP-1 Monoclonal specifically detects TIMP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications
| TIMP-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMP1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7076 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF, ELISA | |
| 63515 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P01033 | |
| TIMP1 | |
| Mouse myeloma cell line NS0-derived recombinant human TIMP-1 Cys24-Ala207 Accession # P01033 | |
| 25 μg | |
| Primary | |
| Detects human TIMP-1 in ELISAs and Western blots. In ELISAs, this antibody does not cross-react with recombinant mouse TIMP-1 or recombinant human TIMP-2. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction