missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TIM-4 ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EH455RBX10
Additional Details : Weight : 2.50000kg
Description
The T cell immunoglobulin and mucin domain containing protein (TIM) family encodes cell surface receptors that are involved in the regulation of T helper (Th) -1 and -2 cell-mediated immunity. TIM-4, which is preferentially expressed on macrophages and dendritic cells, is the natural ligand of TIM-1, and this binding leads to T-cell expansion and cytokine production. Unlike other members of the TIM family, TIM-4 lacks a putative tyrosine phosphorylation signal sequence in its intracellular domain. The TIM-4 gene maps to a locus associated with predisposition to asthma in both mice and humans and with its connection to TIM-1-triggered Th2 responsiveness, may be considered as a candidate disease/predisposition gene for asthma.Specifications
Q96H15 | |
20 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
TIMD4 | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
20 to 5,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
91937 | |
<12% | |
Pre-coated 96 well plate|Standard|Assay Diluent concentrate|Biotinylated Detection Antibody|SAV-HRP|Wash Buffer|Chromogen|Stop Solution|Adhesive Plate Covers | |
10 x 96 Tests | |
Serum, 50 μL|Supernatant, 50 μL|Plasma, 50 μL | |
TIM-4 (TIMD4) | |
4 hr 45 min |