missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TIM-4 ELISA Kit
Human TIM-4 quantitates human TIM-4 in serum, plasma, supernatant.
£540.00 - £2320.00
Specifications
Accession Number | B430010N18Rik; RGD1564516; smuckler; soluble TIM 4; Spleen, mucin-containing, knockout of lymphotoxin protein; sTIM 4; T cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain-containing protein 4; T-cell immunoglobulin mucin receptor 4; T-cell membrane protein 4; Tim4; TIM-4; TIMD4; TIMD-4 |
---|---|
Assay Range | 20 to 5,000 pg/mL |
Assay Sensitivity | 20 pg/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17175239
|
Invitrogen™
EH455RBX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
17144763
|
Invitrogen™
EH455RB |
96 Tests |
£540.00
Pack of 96 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
Description
Human TIM-4 quantitates human TIM-4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human TIM-4.
The T cell immunoglobulin and mucin domain containing protein (TIM) family encodes cell surface receptors that are involved in the regulation of T helper (Th) -1 and -2 cell-mediated immunity. TIM-4, which is preferentially expressed on macrophages and dendritic cells, is the natural ligand of TIM-1, and this binding leads to T-cell expansion and cytokine production. Unlike other members of the TIM family, TIM-4 lacks a putative tyrosine phosphorylation signal sequence in its intracellular domain. The TIM-4 gene maps to a locus associated with predisposition to asthma in both mice and humans and with its connection to TIM-1-triggered Th2 responsiveness, may be considered as a candidate disease/predisposition gene for asthma.Spécification
B430010N18Rik; RGD1564516; smuckler; soluble TIM 4; Spleen, mucin-containing, knockout of lymphotoxin protein; sTIM 4; T cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain-containing protein 4; T-cell immunoglobulin mucin receptor 4; T-cell membrane protein 4; Tim4; TIM-4; TIMD4; TIMD-4 | |
20 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
TIMD4 | |
<10% | |
HRP | |
Serum, 50 μL; Supernatant, 50 μL; Plasma, 50 μL | |
TIM-4 (TIMD4) | |
4 hr 45 min |
20 to 5,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q96H15 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |