missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human TIGAR Recombinant Protein, PeproTech®

Product Code. 17841773
Click to view available options
Quantity:
100 μg
2 x 500 μg
25 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
250µg
25µg
500µg
5µg
Pack of 2
This item is not returnable. View return policy

Product Code. 17841773

Brand: Gibco™ 15014100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

150-14-1MG will be provided as 2 x 500 μg (150-14-500UG). Recombinant Human TIGAR expressed in E.coli is a 29.9 kDa protein containing 269 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene encodes a member of the kelch family of proteins, which are characterized by kelch repeat motifs and a POZ/BTB protein-binding domain. It is thought that kelch repeats are actin binding domains. However, the specific function of this protein has not been determined. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NQ88
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 57103
Molecular Weight (g/mol) 29.9 kDa
Name Human TIGAR
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 9630033F20Rik; AA793651; AI595337; C12orf5; C27H12orf5; C79710; C85509; FR2BP; Fructose-2,6-bisphosphatase TIGAR; fructose-2,6-bisphosphate 2-phosphatase; hypothetical protein LOC502894; LOC502894; probable fructose-2,6-bisphosphatase TIGAR; TIGAR; TP53 induced glycolysis regulatory phosphatase; TP53-induced glycolysis and apoptosis regulator; TP53-induced glycolysis regulatory phosphatase; transactivated by NS3TP2 protein; Trp53 induced glycolysis repulatory phosphatase
Common Name TIGAR
Gene Symbol TIGAR
Conjugate Unconjugated
Recombinant Recombinant
Sequence ARFALTVVRH GETRFNKEKI IQGQGVDEPL SETGFKQAAA AGIFLNNVKF THAFSSDLMR TKQTMHGILE RSKFCKDMTV KYDSRLRERK YGVVEGKALS ELRAMAKAAR EECPVFTPPG GETLDQVKMR GIDFFEFLCQ LILKEADQKE QFSQGSPSNC LETSLAEIFP LGKNHSSKVN SDSGIPGLAA SVLVVSHGAY MRSLFDYFLT DLKCSLPATL SRSELMSVTP NTGMSLFIIN FEEGREVKPT VQCICMNLQD HLNGLTETR
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.