missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Tie-2 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 11 publications
Brand: R&D Systems FAB3131P
This item is not returnable.
View return policy
Description
Tie-2 Monoclonal specifically detects Tie-2 in Human samples. It is validated for Flow Cytometry.
Specifications
| Tie-2 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK, TEK tyrosine kinase, endothelial, Tie2, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7010 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 83715 | |
| Flow Cytometry 10 uL/10^6 cells | |
| AAA61139 | |
| TEK | |
| Mouse myeloma cell line NS0-derived recombinant human Tie-2 Ala23-Lys745 Accession # AAA61139 | |
| 100 Tests | |
| Primary | |
| Detects human Tie-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Tie-1, recombinant mouse Tie-2, or recombinant zebrafish Tie-2 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction