missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Tie-2 Biotinylated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems BAM3313
This item is not returnable.
View return policy
Description
Tie-2 Monoclonal specifically detects Tie-2 in Human samples. It is validated for ELISA Detection (Matched Antibody Pair).
Specifications
| Tie-2 | |
| Monoclonal | |
| LYOPH | |
| ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL | |
| Q02763 | |
| TEK | |
| Mouse myeloma cell line NS0-derived recombinant human Tie-2 Ala23-Lys745 Accession # Q02763 | |
| 250 μg | |
| Primary | |
| Detects human Tie-2 in ELISAs. In ELISAs, this antibody does not cross-react or interfere with recombinant human (rh) Tie-1, recombinant mouse Tie-2, recombinant zebrafish Tie-2, recombinant rat Tie-2, rhAng-1, rhAng-2, or rhAng-4. | |
| Human | |
| Purified |
| ELISA | |
| 83704 | |
| Biotin | |
| Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
| angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK, TEK tyrosine kinase, endothelial, Tie2, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7010 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction