missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Thymopoietin Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF843
This item is not returnable.
View return policy
Description
Thymopoietin/LAP2 Polyclonal specifically detects Thymopoietin/LAP2 in Human samples. It is validated for Western Blot.
Specifications
| Thymopoietin/LAP2 | |
| Polyclonal | |
| Unconjugated | |
| CMD1T, lamina-associated polypeptide 2, LAP2PRO0868, LEM domain containing 4, LEMD4, MGC61508, thymopoietin, Thymopoietin isoform alpha, Thymopoietin, isoforms beta/gamma, Thymopoietin-related peptide isoform alpha, Thymopoietin-related peptide isoforms beta/gamma, TMPO, TP, TP alpha, TP beta/gamma, TPRP isoform alpha, TPRP isoforms beta/gamma | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7112 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| TMPO | |
| E. coli-derived recombinant human Thymopoietin Pro2-Gln187 | |
| 100 μg | |
| Primary | |
| Detects human Thymopoietin in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction