missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Thymidine Kinase 1 Antibody, R&D Systems™

Sheep Antibody
£157.00 - £410.00
Specifications
| Antigen | Thymidine Kinase 1 |
|---|---|
| Dilution | Simple Western 20 ug/mL, Immunohistochemistry 1-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18725536
![]() |
R&D Systems
AF8180-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18719246
![]() |
R&D Systems
AF8180-100 |
100 μg |
£410.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thymidine Kinase 1 specifically detects Thymidine Kinase 1 in Human samples. It is validated for Simple Western, Immunohistochemistry.Specifications
| Thymidine Kinase 1 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Sheep | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EC 2.7.1.21, thymidine kinase 1 soluble isoform, thymidine kinase 1, soluble, thymidine kinase, cytosolic, thymidine kinase-1, TK1, TK2 | |
| E. coli-derived human Thymidine Kinase 1 protein, Ser2-Asn234, Accession No. P04183 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Thymidine Kinase 1 Protein in direct ELISAs. |
| Simple Western 20 ug/mL, Immunohistochemistry 1-25 ug/mL | |
| Polyclonal | |
| Lyophilized | |
| Human | |
| P04183 | |
| 7083.0 | |
| IgG | |
| Antigen Affinity-purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title