missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Thrombospondin-4 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB2390
This item is not returnable.
View return policy
Description
Thrombospondin-4 Monoclonal specifically detects Thrombospondin-4 in Human samples. It is validated for Western Blot.
Specifications
| Thrombospondin-4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P35443 | |
| THBS4 | |
| Mouse myeloma cell line NS0-derived recombinant human Thrombospondin-4 Ala22-Asn961 (Pro276Ala, Ala420Val) Accession # P35443 | |
| 100 μg | |
| Primary | |
| Detects human Thrombospondin-4 in direct ELISAs and Western blots. Does not cross-react with recombinant human Thrombospondin-1, -2, or -3. | |
| Human | |
| Purified |
| Western Blot | |
| 276523 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| THBS4, thrombospondin 4, thrombospondin-4, Thrombospondin4, TSP4, TSP-4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7060 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction