Learn More
Human Thrombospondin-2 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand:  R&D Systems AF1635
 View more versions of this product
Product Code. 18038261
  • £506.00 / 100µg
Description
Thrombospondin-2 Polyclonal specifically detects Thrombospondin-2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thrombospondin-2
Polyclonal
Unconjugated
P35442
THBS2
Mouse myeloma cell line NS0-derived recombinant human Thrombospondin-2 Gly19-Ile1172 Accession # P35442
100 μg
Primary
Detects human Thrombospondin-2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Thrombospondin-1 and rhThrombospondin-4 is observed.
Human
IgG
Western Blot
LYOPH
Western Blot 0.1 ug/mL
THBS2, thrombospondin 2, Thrombospondin2, TSP-2, TSP2thrombospondin-2
Goat
Affinity Purified
RUO
7058
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.